Ethyl dichlorofluoroacetate 96%

Supplier: APOLLO SCIENTIFIC
Danger

PC450268-1G PC450268-5G
77415-518EA 69.28 USD
77415-518 77415-520
Ethyl dichlorofluoroacetate 96%
Ethyl dichlorofluoroacetate
Formula: C₄H₅Cl₂FO₂
MW: 174.986 g/mol
Boiling Pt: 130…132 °C
Density: 1.307 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013664
CAS Number: 383-61-9
UN: 3265
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR