2-Fluoro-5-(trifluoromethyl)pyridine 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC4374-100G PC4374-25G
77415-288EA 58.08 USD
77415-288 77415-290
2-Fluoro-5-(trifluoromethyl)pyridine 97%
2-Fluoro-5-(trifluoromethyl)pyridine
Formula: C₆H₃F₄N
MW: 165.09 g/mol
Boiling Pt: 118…122 °C
Density: 1.37 g/cm³ (25 °C)
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD07437945
CAS Number: 69045-82-5
EINECS: 400-290-2
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR