3-Fluoro-5-(trifluoromethyl)benzonitrile 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC4373I-100G PC4373I-25G PC4373I-5G
77415-276EA 158.78 USD
77415-276 77415-278 77415-280
3-Fluoro-5-(trifluoromethyl)benzonitrile 97%
3-Fluoro-5-(trifluoromethyl)benzonitrile
Formula: C₈H₃F₄N
Density: 1.35 g/cm³ (25 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00061282
CAS Number: 149793-69-1
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR