5-Fluoro-o-tolylisothiocyanate 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Methyl-5-fluorophenylisothiocyanate, 5-Fluoro-2-methylphenyl Isothiocyanate, Isothiocyanic Acid 5-Fluoro-o-tolyl Ester

PC3823W-100G PC3823W-25G PC3823W-5G
77414-946EA 309.96 USD
77414-946 77414-948 77414-950
5-Fluoro-o-tolylisothiocyanate 97%
5-Fluoro-o-tolylisothiocyanate
Formula: C₈H₆FNS
MW: 167.20 g/mol
Boiling Pt: 101 °C/20 mmHg
Storage Temperature: Ambient
MDL Number: MFCD00046800
CAS Number: 175205-39-7
EINECS: 223-167-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR