5-Fluoro-o-toluonitrile 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC3823C-100G PC3823C-10G PC3823C-25G
77414-934EA 108 USD
77414-934 77414-936 77414-938
5-Fluoro-o-toluonitrile 98%
5-Fluoro-o-toluonitrile
Formula: C₈H₆FN
Melting Pt: 42…45 °C
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00042295
CAS Number: 77532-79-7
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR