5-Fluoro-2'-deoxyuridine (floxuridine) 99%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC3740-1G
PC3740-25G
PC3740-5G
77414-862EA
32.6
USD
77414-862
77414-864
77414-866
5-Fluoro-2'-deoxyuridine (floxuridine) 99%
5-Fluoro-2'-deoxyuridine (floxuridine)
Formula:
C₉H₁₁FN₂O₅ Melting Pt: 148…151 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006530 CAS Number: 50-91-9 EINECS: 200-072-5 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...