3-Fluorobenzoyl chloride 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC3620-100G PC3620-25G
77414-786EA 45.84 USD
77414-786 77414-788
3-Fluorobenzoyl chloride 98%
3-Fluorobenzoyl chloride
Formula: C₇H₄ClFO
Boiling Pt: 189 °C
Melting Pt: –30 °C
Density: 1.32 g/cm³ (25 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00000670
CAS Number: 1711-07-5
EINECS: 216-977-3
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR