3-Bromo-4-fluorophenylacetylene 96%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
PC56838-1G
PC56838-250MG
77416-892EA
544.04
USD
77416-892
77416-894
3-Bromo-4-fluorophenylacetylene 96%
3-Bromo-4-fluorophenylacetylene
Formula:
C₈H₄BrF Storage Temperature: Refrigerator |
MDL Number:
MFCD26395447 CAS Number: 918487-39-5 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...