6-Bromo-3-(difluoromethyl)-1H-indole, N-BOC protected

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: tert-butyl 6-bromo-3-(difluoromethyl)indole-1-carboxylate

PC56795-1G PC56795-500MG PC56795-100MG
77416-874EA 2659.46 USD
77416-874 77416-876 77416-872
6-Bromo-3-(difluoromethyl)-1H-indole, N-BOC protected
6-Bromo-3-(difluoromethyl)-1H-indole, N-BOC protected
Formula: C₁₄H₁₄BrF₂NO₂
MW: 346.167 g/mol
Storage Temperature: Ambient
CAS Number: 2237235-37-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR