(4-Chlorosulfonyl-2-fluorophenoxy)acetic acid methyl ester
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC53525-100MG
PC53525-250MG
77416-708EA
139.58
USD
77416-708
77416-710
(4-Chlorosulfonyl-2-fluorophenoxy)acetic acid methyl ester
(4-Chlorosulfonyl-2-fluorophenoxy)acetic acid methyl ester
Formula:
C₉H₈ClFO₅S MW: 282.673 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18838355 CAS Number: 1291488-79-3 UN: 1759 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...