4-Chloro-3-iodobenzotrifluoride, Technical Grade

Supplier: APOLLO SCIENTIFIC
Danger

PC53433-100G PC53433-1KG PC53433-500G
77416-694EA 44.26 USD
77416-694 77416-696 77416-698
4-Chloro-3-iodobenzotrifluoride, Technical Grade
4-Chloro-3-iodobenzotrifluoride
Formula: C₇H₃ClF₃I
Storage Temperature: Refrigerator
MDL Number: MFCD00001037
CAS Number: 672-57-1
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR