2-Fluoro-α-methyl-4-biphenylacetic acid 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Ansaid, 2-(2-Fluoro-[1,1'-biphenyl]-4-yl)propanoic acid, Flurbiprofen, Froben, Antadys

PC5245-100G PC5245-25G PC5245-5G
77416-544EA 481.9 USD
77416-544 77416-546 77416-548
2-Fluoro-α-methyl-4-biphenylacetic acid 98%
2-Fluoro-α-methyl-4-biphenylacetic acid
Formula: C₁₅H₁₃FO₂
MW: 244.265 g/mol
Melting Pt: 110…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00079303
CAS Number: 5104-49-4
EINECS: 225-827-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR