Ethyl 2-bromo-4,4-difluoro-3-oxobutanoate, Technical Grade

Supplier: APOLLO SCIENTIFIC
Warning

PC52197-1G PC52197-250MG
77416-534EA 406.5 USD
77416-534 77416-536
Ethyl 2-bromo-4,4-difluoro-3-oxobutanoate, Technical Grade
Ethyl 2-bromo-4,4-difluoro-3-oxobutanoate
Formula: C₆H₇BrF₂O₃
MW: 245.019 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27940538
CAS Number: 660840-16-4
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR