Methyl trifluoropyruvate 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC5185E-25G PC5185E-5G
77416-466EA 31.6 USD
77416-466 77416-468
Methyl trifluoropyruvate 95%
Methyl trifluoropyruvate
Formula: C₄H₃F₃O₃
Boiling Pt: 84…86 °C
Density: 1.529 g/cm³ (25 °C)
Flash Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD00114936
CAS Number: 13089-11-7
UN: 3272
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR