3-Fluoropyridine 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC4201-100G PC4201-500G
77415-200EA 83.54 USD
77415-200 77415-202
3-Fluoropyridine 98%
3-Fluoropyridine

Tetrahedron, 39, 2009 (1983): with LDA lithiation is at the 2-position initially, however with longer reaction times the major product is the 4-litho derivative. Ask for other refs.

Formula: C₅H₄FN
MW: 97.09 g/mol
Boiling Pt: 106…108 °C
Density: 1.13 g/cm³ (25 °C)
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00006374
CAS Number: 372-47-4
EINECS: 206-755-4
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR