1-(4-Fluorobenzyl)homopiperazine 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC4089-10G PC4089-1G PC4089-5G
77415-118EA 151.8 USD
77415-118 77415-120 77415-122
1-(4-Fluorobenzyl)homopiperazine 95%
1-(4-Fluorobenzyl)homopiperazine
Formula: C₁₂H₁₇FN₂
MW: 208.28 g/mol
Boiling Pt: 112 °C/0.5 mmHg
Storage Temperature: Ambient
MDL Number: MFCD00816588
CAS Number: 76141-89-4
UN: 3267
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR