2,6-Difluoro-m-toluidine 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,6-Difluoro-3-methylaniline

PC51074-10G PC51074-1G PC51074-5G
77416-436EA 289.34 USD
77416-436 77416-438 77416-440
2,6-Difluoro-m-toluidine 98%
2,6-Difluoro-m-toluidine
Formula: C₇H₇F₂N
MW: 143.14
Storage Temperature: Refrigerator
MDL Number: MFCD06660182
CAS Number: 144851-63-8
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR