2-Mercapto-5-(trifluoromethyl)pyridine 95%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
5-(Trifluoromethyl)-2(1H)-pyridinethione, 5-(Trifluoromethyl)-2-pyridinethiol, 5-(Trifluoromethyl)pyridine-2(1H)-thione
PC5043E-100G
PC5043E-25G
PC5043E-5G
77416-368EA
329.08
USD
77416-368
77416-370
77416-372
2-Mercapto-5-(trifluoromethyl)pyridine 95%
2-Mercapto-5-(trifluoromethyl)pyridine
Formula:
C₆H₄F₃NS MW: 179.16 g/mol Melting Pt: 147…160 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00128893 CAS Number: 76041-72-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...