4-Bromo-3,5-difluorobenzenesulfonyl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-Bromo-3,5-difluorobenzene-1-sulphonyl chloride

PC4965-1G PC4965-25G PC4965-5G
77415-948EA 21.4 USD
77415-948 77415-950 77415-952
4-Bromo-3,5-difluorobenzenesulfonyl chloride 97%
4-Bromo-3,5-difluorobenzenesulfonyl chloride
Formula: C₆H₂BrClF₂O₂S
MW: 291.498 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03788548
CAS Number: 518057-63-1
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR