2-Fluoropropenoic acid 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC4962-1G PC4962-5G
77415-938EA 54 USD
77415-938 77415-940
2-Fluoropropenoic acid 97%
2-Fluoropropenoic acid
Formula: C₃H₃FO₂
Melting Pt: 51…52 °C
Storage Temperature: Freezer
MDL Number: MFCD03424474
CAS Number: 430-99-9
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR