1,1,1,2,2,3,3-Heptafluoro-3-[(trifluorovinyl)oxy]propane 97%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
Perfluoropropoxyethylene, (Heptafluoropropoxy)trifluoroethene, Perfluoro(propyl vnyl ether)
PC4513L-100G
PC4513L-25G
77415-600EA
391.22
USD
77415-600
77415-602
1,1,1,2,2,3,3-Heptafluoro-3-[(trifluorovinyl)oxy]propane 97%
1,1,1,2,2,3,3-Heptafluoro-3-[(trifluorovinyl)oxy]propane
Useful intermediate and monomer; the co-polymer with tetrafluoroethylene is sold as Teflon PFA.
Formula:
C₅F₁₀O MW: 266.04 g/mol Boiling Pt: 35…36 °C Melting Pt: –70 °C Density: 1.53 g/cm³ (25 °C) Flash Pt: –20 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00191577 CAS Number: 1623-05-8 EINECS: 216-600-2 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...