2-Fluoro-3-methoxybenzylamine

Supplier: APOLLO SCIENTIFIC
Danger

PC408258-1G PC408258-5G
77415-112EA 101.88 USD
77415-112 77415-114
2-Fluoro-3-methoxybenzylamine
2-Fluoro-3-methoxybenzylamine
Formula: C₈H₁₀FNO
Storage Temperature: Ambient
MDL Number: MFCD11045854
CAS Number: 93071-81-9
UN: 3267
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR