Ethyl-2-methyl-4,4,4-trifluoroacetoacetate 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC3245D-100G PC3245D-25G PC3245D-5G
77414-598EA 330.1 USD
77414-598 77414-600 77414-602
Ethyl-2-methyl-4,4,4-trifluoroacetoacetate 97%
Ethyl-2-methyl-4,4,4-trifluoroacetoacetate
Formula: C₇H₉F₃O₃
Boiling Pt: 144…146 °C
Density: 1.258 g/cm³ (25 °C)
Flash Pt: 47 °C
Storage Temperature: Ambient
MDL Number: MFCD00190635
CAS Number: 344-00-3
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR