11H-Perfluoroundecanoic acid 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC3148-1G PC3148-5G
77414-522EA 69.28 USD
77414-522 77414-524
11H-Perfluoroundecanoic acid 97%
11H-Perfluoroundecanoic acid
Formula: C₁₁H₂F₂₀O₂
Melting Pt: 100…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00042431
CAS Number: 1765-48-6
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR