3-Fluorobenzyl isothiocyanate 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC3721M-1G PC3721M-250MG
77414-828EA 61.12 USD
77414-828 77414-830
3-Fluorobenzyl isothiocyanate 97%
3-Fluorobenzyl isothiocyanate
Formula: C₈H₆FNS
MW: 167.21 g/mol
Boiling Pt: 138 °C/16 mmHg
Density: 1.22 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00060406
CAS Number: 63351-94-0
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR