α-Bromo-3-fluorotoulene 96%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: α-Bromo-3-fluorotoluene, α-Bromo-m-fluorotoulene, alpha-Bromo-m-fluorotoulene, 1-(Bromomethyl)-3-fluorobenzene, m-Fluorobenzyl bromide, 3-Fluorobenzyl bromide

PC3694-100G PC3694-25G
77414-820EA 41.78 USD
77414-820 77414-822
α-Bromo-3-fluorotoulene 96%
α-Bromo-3-fluorotoulene
Formula: C₇H₆BrF
Boiling Pt: 88 °C/20 mmHg
Density: 1.541 g/cm³ (25 °C)
Flash Pt: 61 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000340
CAS Number: 456-41-7
EINECS: 207-263-2
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR