Ethyl-4,4,4-trifluoro-3-(trifluoromethyl)butyrate 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Ethyl (2H-perfluoroprop-2-yl)acetate

PC3289E-1G PC3289E-5G
77414-668EA 48.9 USD
77414-668 77414-670
Ethyl-4,4,4-trifluoro-3-(trifluoromethyl)butyrate 97%
Ethyl-4,4,4-trifluoro-3-(trifluoromethyl)butyrate
Formula: C₇H₈F₆O₂
Flash Pt: 15 °C
Storage Temperature: Ambient
MDL Number: MFCD00236608
CAS Number: 17327-34-3
UN: 3272
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR