Ethyl-4,4,4-trifluoroacetoacetate 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Ethyl trifluoroacetoacetate, Ethyl 4,4,4-trifluoro-3-oxobutanoate

PC3280-100G PC3280-25G PC3280-500G
77414-650EA 28.52 USD
77414-650 77414-652 77414-654
Ethyl-4,4,4-trifluoroacetoacetate 95%
Ethyl-4,4,4-trifluoroacetoacetate
Formula: C₆H₇F₃O₃
MW: 184.11 g/mol
Boiling Pt: 70 °C/100 mmHg
Melting Pt: –39 °C
Density: 1.259 g/cm³ (25 °C)
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00000424
CAS Number: 372-31-6
EINECS: 206-750-7
UN: 3272
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR