(2R)-(+)-3,3,3-Trifluoro-1,2-propenoxide

Supplier: APOLLO SCIENTIFIC

PC32742-1G PC32742-250MG PC32742-5G
77414-634EA 303.6 USD
77414-634 77414-636 77414-638
(2R)-(+)-3,3,3-Trifluoro-1,2-propenoxide
(2R)-(+)-3,3,3-Trifluoro-1,2-propenoxide
Formula: C₃H₃F₃O
Boiling Pt: 25…32 °C
Melting Pt: –80 °C
Density: 1.294 g/cm³ (25 °C)
Flash Pt: –26 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08276876
CAS Number: 143142-90-9
UN: 1993
ADR: 3,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR