3-Fluoro-4-nitropyridine 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC3238-1G PC3238-250MG PC3238-5G
77414-586EA 144.1 USD
77414-586 77414-588 77414-590
3-Fluoro-4-nitropyridine 95%
3-Fluoro-4-nitropyridine
Formula: C₅H₃FN₂O₂
Boiling Pt: 62…64 °C/5 mmHg
Flash Pt: 93 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04112517
CAS Number: 13505-01-6
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR