Ethyl chlorofluoroacetate 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Ethylchlorofluoroacetate

PC3180-100G PC3180-25G
77414-558EA 225.88 USD
77414-558 77414-560 77414-562
Ethyl chlorofluoroacetate 97%
Ethyl chlorofluoroacetate
Formula: C₄H₆ClFO₂
Boiling Pt: 128…130 °C
Density: 1.225 g/cm³ (25 °C)
Flash Pt: 44 °C
Storage Temperature: Ambient
MDL Number: MFCD00039333
CAS Number: 401-56-9
EINECS: 206-930-5
UN: 2920
ADR: 8+3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR