2,4-Bis(trifluoromethyl)benzyl chloride 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC3105D-100G PC3105D-25G PC3105D-5G
77414-486EA 1322.42 USD
77414-486 77414-488 77414-490
2,4-Bis(trifluoromethyl)benzyl chloride 98%
2,4-Bis(trifluoromethyl)benzyl chloride
Formula: C₉H₅ClF₆
Storage Temperature: Ambient
MDL Number: MFCD00083537
CAS Number: 195136-46-0
UN: 1760
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR