3,6-Dichloro-2-fluorobenzyl bromide 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC302703-1G PC302703-5G
77414-248EA 49.92 USD
77414-248 77414-250
3,6-Dichloro-2-fluorobenzyl bromide 98%
3,6-Dichloro-2-fluorobenzyl bromide
Formula: C₇H₄BrCl₂F
Melting Pt: 42…45 °C
Storage Temperature: Ambient
MDL Number: MFCD09025389
CAS Number: 916420-69-4
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR