2-Fluorophenyl isocyanate 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-Fluoro-2-isocyanatobenzene

PC4164-1G PC4164-5G
77415-174EA 33.62 USD
77415-174 77415-176
2-Fluorophenyl isocyanate 98%
2-Fluorophenyl isocyanate

  • May form small amount of sediment.

Formula: C₇H₄FNO
MW: 137.11 g/mol
Boiling Pt: 56…58 °C/10 mmHg
Density: 1.222 g/cm³ (25 °C)
Flash Pt: 50 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001996
CAS Number: 16744-98-2
EINECS: 240-804-0
UN: 3080
ADR: 6.1+3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR