2,2,2-Trifluoroethyl methanesulfonate 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC3889-5G PC3889-1G PC3889-25G
77414-994EA 69.28 USD
77414-994 77414-990 77414-992
2,2,2-Trifluoroethyl methanesulfonate 98%
2,2,2-Trifluoroethyl methanesulfonate
Formula: C₃H₅F₃O₃S
Density: 1.513 g/cm³ (25 °C)
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00025058
CAS Number: 25236-64-0
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR