3'-(Trifluoromethylthio)acetophenone 94%

Supplier: APOLLO SCIENTIFIC
Warning

PC3821-1G PC3821-25G PC3821-5G
77414-922EA 102.5 USD
77414-922 77414-924 77414-926
3'-(Trifluoromethylthio)acetophenone 94%
3'-(Trifluoromethylthio)acetophenone
Formula: C₉H₇F₃OS
Storage Temperature: Refrigerator
MDL Number: MFCD00236331
CAS Number: 56773-33-2
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR