2-Fluorobiphenyl 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Fluoro-1,1'-biphenyl, PFB No. 1

PC3724-100G PC3724-25G PC3724-5G
77414-838EA 75.4 USD
77414-838 77414-840 77414-842
2-Fluorobiphenyl 98%
2-Fluorobiphenyl
Formula: C₁₂H₉F
MW: 172.2 g/mol
Boiling Pt: 248 °C
Melting Pt: 71…76 °C
Density: 1.245 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000317
CAS Number: 321-60-8
EINECS: 206-290-7
UN: 3077
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR