4-Fluorobenzenesulfonyl chloride 95%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC3490-500G
PC3490-50G
PC3490-100G
77414-754EA
166.06
USD
77414-754
77414-756
77414-752
4-Fluorobenzenesulfonyl chloride 95%
4-Fluorobenzenesulphonyl chloride
Formula:
C₆H₄ClFO₂S Boiling Pt: 95…96 °C/2 mmHg Melting Pt: 29…31 °C Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007438 CAS Number: 349-88-2 EINECS: 206-493-0 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...