Ethyl-4,4,4-trifluoro-3-(trifluoromethyl)crotonate 95%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
PC3289G-25G
PC3289G-5G
77414-674EA
653.52
USD
77414-674
77414-676
77414-672
Ethyl-4,4,4-trifluoro-3-(trifluoromethyl)crotonate 95%
Ethyl-4,4,4-trifluoro-3-(trifluoromethyl)crotonate
Formula:
C₇H₆F₆O₂ Boiling Pt: 127…128 °C Density: 1.3525 g/cm³ (25 °C) Flash Pt: 46 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041517 CAS Number: 1513-60-6 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...