4-Fluoro-2-methylbenzyl bromide 98%
Supplier: APOLLO SCIENTIFIC
Synonyms:
4-Fluoro-2-methylbenzyl Bromide
PC302785-1G
PC302785-25G
PC302785-5G
77414-264EA
32.6
USD
77414-264
77414-266
77414-268
4-Fluoro-2-methylbenzyl bromide 98%
4-Fluoro-2-methylbenzyl bromide
Formula:
C₈H₈BrF MW: 203.05 g/mol Boiling Pt: 142 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03701059 CAS Number: 862539-91-1 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...