2-Fluoro-4-(pentafluorothio)benzyl bromide

Supplier: APOLLO SCIENTIFIC
Danger

PC302452-1G PC302452-5G
77414-186EA 242.7 USD
77414-186 77414-188
2-Fluoro-4-(pentafluorothio)benzyl bromide
2-Fluoro-4-(pentafluorothio)benzyl bromide
Formula: C₇H₅BrF₆S
Storage Temperature: Ambient
MDL Number: MFCD16652507
CAS Number: 1240257-17-3
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR