2-(Difluoromethoxy)-4-fluorobenzyl bromide 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC302419-10G PC302419-1G PC302419-5G
77414-176EA 842.56 USD
77414-176 77414-178 77414-180
2-(Difluoromethoxy)-4-fluorobenzyl bromide 98%
2-(Difluoromethoxy)-4-fluorobenzyl bromide
Formula: C₈H₆BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD09832363
CAS Number: 1017779-42-8
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR