2-Chloro-4,5-difluorophenylacetonitrile 98%

Supplier: APOLLO SCIENTIFIC

PC302023-1G PC302023-25G PC302023-5G
77414-098EA 99.84 USD
77414-098 77414-100 77414-102
2-Chloro-4,5-difluorophenylacetonitrile 98%
2-Chloro-4,5-difluorophenylacetonitrile
Formula: C₈H₄ClF₂N
MW: 187.574 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660213
CAS Number: 874285-22-0
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR