2,5-Difluoro-4-morpholin-4-ylbenzoic acid

Supplier: APOLLO SCIENTIFIC

PC300787-100MG PC300787-1G
77414-030EA 48.9 USD
77414-030 77414-032
2,5-Difluoro-4-morpholin-4-ylbenzoic acid
2,5-Difluoro-4-morpholin-4-ylbenzoic acid
Formula: C₁₁H₁₁F₂NO₃
MW: 243.207 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28103348
CAS Number: 1430223-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR