2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}benzoic acid

Supplier: APOLLO SCIENTIFIC

PC300779-1G PC300779-5G
77414-022EA 176.26 USD
77414-022 77414-024
2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}benzoic acid
2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}benzoic acid
Formula: C₁₃H₇ClF₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD03644663
CAS Number: 852952-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR