Buten-1-ylsulfur pentafluoride

Supplier: APOLLO SCIENTIFIC
Warning

PC450557-1G PC450557-250MG
77415-564EA 655.1 USD
77415-564 77415-566
Buten-1-ylsulfur pentafluoride
Buten-1-ylsulfur pentafluoride
Formula: C₄H₇F₅S
Storage Temperature: Ambient
MDL Number: MFCD31561017
CAS Number: 2149605-96-7
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR