(2,2-Difluoroethoxy)benzene

Supplier: APOLLO SCIENTIFIC
Warning

PC450195-1G PC450195-250MG PC450195-5G
77415-486EA 268.96 USD
77415-486 77415-488 77415-490
(2,2-Difluoroethoxy)benzene
(2,2-Difluoroethoxy)benzene
Formula: C₈H₈F₂O
MW: 158.145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19442073
CAS Number: 1309602-18-3
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR