2,2,3,3,3-Pentafluoropropyl butyrate

Supplier: APOLLO SCIENTIFIC
Warning

PC450190-1G PC450190-5G
77415-476EA 93.74 USD
77415-476 77415-478
2,2,3,3,3-Pentafluoropropyl butyrate
2,2,3,3,3-Pentafluoropropyl butyrate
Formula: C₇H₉F₅O₂
MW: 220.137 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22123979
CAS Number: 950191-45-4
UN: 3272
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR