Heptafluorobutyric acid 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Perfluorobutyric acid, HFBA, Heptafluorobutanoic acid

PC4450-100G PC4450-250G PC4450-25G
77415-410EA 96.78 USD
77415-410 77415-412 77415-414
Heptafluorobutyric acid 98%
Heptafluorobutyric acid

Biochemistry, 20, 4530 (1981): anion-pairing agent for reverse phase HPLC purification of proteins & peptides.

Formula: C₄HF₇O₂
MW: 214.04 g/mol
Boiling Pt: 119…121 °C
Melting Pt: –18 °C
Density: 1.645 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004171
CAS Number: 375-22-4
EINECS: 206-786-3
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR