1,3-difluoro-2-methyl-4-nitrobenzene 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC400731-10G PC400731-1G PC400731-5G
77415-050EA 74.38 USD
77415-050 77415-052 77415-054
1,3-difluoro-2-methyl-4-nitrobenzene 97%
1,3-difluoro-2-methyl-4-nitrobenzene
Formula: C₇H₅F₂NO₂
Melting Pt: 35…38 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11519030
CAS Number: 79562-49-5
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR